Order Entry
Puerto Rico
ContactUsLinkComponent
Potassium hydroxide 0.5 N in ethanol, BAKER ANALYZED® volumetric solution, J.T.Baker®
Flammable
Corrosive
Irritant
Health
Potassium hydroxide 0.5 N in ethanol, BAKER ANALYZED® volumetric solution, J.T.Baker®
Catalog # JT5644-2
CAS Number:  
MOC:  Non-RegulatedCertificates
undefined
Flammable
Corrosive
Irritant
Health
Potassium hydroxide 0.5 N in ethanol, BAKER ANALYZED® volumetric solution, J.T.Baker®
Catalog # JT5644-2
Supplier Number:  5644-02
CAS Number:  
MOC:  Non-RegulatedCertificates

Specifications

  • Pack type:
    PE bottle, narrow mouth
  • Size:
    1 L
  • Cat. No.:
    JT5644-2
  • Supplier no.:
    5644-02

Specifications

Specification Test Results

  • Normality:
    0.495 - 0.505
  • Appearance:
    Passes Test

Specification Test Results

About this item

 :
 :56.11 g/mol
ADR:3,II
 :Management of Change (MOC) category = N
Flash Pt:13.00 °C
EINECS:215-181-3
UN:2924
 :GHS02
 :GHS05
 :GHS07
 :GHS08
Storage Temperature:Ambient
CAS Number:1310-58-3
Formula:KOH
MDL Number:MFCD00003553