ContactUsLinkComponent
SP Bel-Art Clavies™ Bag Clips, Bel-Art Products, a part of SP
SP Bel-Art Clavies™ Bag Clips, Bel-Art Products, a part of SP
Catalog # 12000-072
undefined
SP Bel-Art Clavies™ Bag Clips, Bel-Art Products, a part of SP
Catalog # 12000-072
Supplier Number:  F13190-0100
Specifications
  • Cat. No.:
    12000-072
  • W×D×H:
    11 mm (⁷/₁₆")
  • Supplier no.:
    131900100
Specifications
About this item
Stiff plastic design provides secure closure for autoclave bags yet allows release of internal pressure that develops during autoclave cycle.
  • Disposable clip is easily closed and opened with one hand
  • Secure closure
Suitable for benchtop or small autoclave bags up to 48.3×58.4 cm (19×23"); use with larger bags is not recommended.