Round-Bottom Three-Neck Flasks, Indented Morton Type, Ace Glass
Supplier: Ace Glass
Certificates
About this item
Round-bottom flasks with angled side necks and [ST] 14/20 outer joints
Indents allow more complete agitation.
Round-bottom flasks with angled side necks and [ST] 14/20 outer joints
Indents allow more complete agitation.