
Sodium hydroxide 20% (w/v) in aqueous solution (200 g/L)
Supplier: Ricca Chemical
Certificates
About this item
CAS Number:1310-73-2
:
Formula:NaOH
MDL Number:MFCD00003548
:40 g/mol
Boiling Pt:119…135 °C (1013 hPa)
Melting Pt:9…10 °C
Density:1.53 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-185-5
UN:1824
ADR:8,II
:GHS05
Specification Test Results
- Appearance:Clear, colorless, odorless
- Assay at 20 °C (traceable to NIST SRM 723):20.00 ± 0.04 % (w/v)
Specification Test Results
Related Information
Certificates
Search by Lot Numberghs_7270
SDS
VWR GHS Compliance Letter (US)