

Sodium hydroxide 5% (w/v) in aqueous solution (50 g/L)
Supplier: Ricca Chemical
Certificates
About this item
CAS Number:1310-73-2
:
Formula:NaOH
MDL Number:MFCD00003548
:40 g/mol
Boiling Pt:~100 °C
Melting Pt:~0 °C
Density:1.53 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-185-5
UN:1824
ADR:8,III
:GHS05
:GHS08
Specification Test Results
- Appearance:Colorless liquid
- Assay (vs. Sulfuric Acid/Phenolphthalein):4.96 - 5.04 % (w/v)
Specification Test Results
Related Information
Certificates
Search by Lot Numberghs_R7250000
SDS
VWR GHS Compliance Letter (US)