



Potassium hydroxide 0.5 N in methanol, BAKER ANALYZED® volumetric solution, J.T.Baker®
Catalog # JT5631-2
Supplier: AVANTOR PERFORMANCE MATERIALS US
CAS Number:
Specifications
- Pack type:PE bottle, narrow mouth
- Size:1 L
- Cat. No.:JT5631-2
- Supplier no.:5631-02
Specifications
Specification Test Results
- Normality:0.497 - 0.503
- Chloride (Cl):≤10 ppm
- Nitrogen Compounds (as N):≤1 ppm
- ACS - Heavy Metals (as Pb):≤1 ppm
- Trace Impurities - Iron (Fe):≤0.5 ppm
Specification Test Results
About this item
:
:56.11 g/mol
Density:0.80 g/cm³
UN:3286
ADR:3,II
:Management of Change (MOC) category = N
EINECS:215-181-3
:GHS02
:GHS06
:GHS05
:GHS08
Storage Temperature:Ambient
CAS Number:1310-58-3
Formula:KOH
MDL Number:MFCD00003553