ContactUsLinkComponent
Zero Charge® Mat and Table Top Cleaner, Techspray™
Zero Charge® Mat and Table Top Cleaner, Techspray™
Catalog # 58542-414
Supplier:  ITW Chemtronics
undefined
Zero Charge® Mat and Table Top Cleaner, Techspray™
Catalog # 58542-414
Supplier:  ITW Chemtronics
Supplier Number:  1733-G
Specifications
  • Size:
    3.78 L (1 gal.)
  • Environmentally Preferable:
  • Cat. No.:
    58542-414
  • Packaging:
    Refill Jug
  • Supplier no.:
    1733-G
Specifications
About this item
A neutral pH cleaner for general use in production areas, this product removes insulating contaminants such as flux residues, oils, greases, nicotine, dirt, and grime
It eliminates charge while leaving an antistatic agent that helps prevent static charge build-up. Use on conductive mats, table tops, computer and soldering stations, telecommunication equipment, floor mats, electronic instrumentation, and more. Nonabrasive, noncorrosive, nonflammable; will not scratch substrate. Contains no ammonia.