
Sodium hydroxide 95.0-100.5%, pellets NF, FCC, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIALS US
MOC: Regulated
About this item
CAS Number:1310-73-2
:
Formula:NaOH
MDL Number:MFCD00003548
:40 g/mol
Merck Index:14,08627
Boiling Pt:1390 °C (1013 hPa)
Melting Pt:323 °C
Density:2.13 g/cm³ (20 °C)
UN:1823
ADR:8,II
:Management of Change (MOC) category = R
Storage Temperature:Ambient
EINECS:215-185-5
REACH:01-2119457892-27
:GHS05
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specification Test Results
- NF - Total Alkali:95.0 - 100.5 %
- NF - Content of Sodium:54.0 - 59.8 %
- NF - Appearance (white hygroscopic pellets):Passes Test
- NF - Insoluble Substances and Organic Matter:Passes Test
- NF - Identification A (Sodium):Passes Test
- NF - Identification B (pH):≥11.0
- NF - Potassium (K):≤0.5 %
- NF - Sodium Carbonate (Na₂CO₃):≤3.0 %
- FCC - Assay (Total alkali as NaOH) (by acidimetry):95.0 - 100.5 %
- FCC - Identification:Passes Test
- FCC - Insoluble Substances and Organic Matter:Passes Test
- FCC - Lead (Pb):≤2 mg/kg
- FCC - Mercury (Hg):≤0.1 mg/kg
- FCC - Arsenic (As):≤3 mg/kg
- FCC - Sodium Carbonate (Na₂CO₃):≤3.0 %
Specification Test Results
Related Information
Certificates
Search by Lot Number7680 Product Specifications VWR GHS Compliance Letter (US)
Additional Product Details
Avantor Portfolio of Chemicals
Product Literature
Macron 7680.pdf
Datasheet
sds_MACR7680
SDS
spec_MK768004 spec_MK768028 spec_MK768088