Standard TouchMaster Cotton Lisle Inspection Gloves, Magid
The Magid's TouchMaster gloves are a 100% cotton, two piece lisle inspection glove constructed of cool and breathable cotton. They are form fitting, lightweight and cut and sewn for the most comfortable fit.
- Cut and sewn pattern
- Color-coded sizes for easy sorting
- Breathable lightweight fabric
This glove can be worn alone to protect delicate products from smudges, finger prints or skin oils, or it can be used as a liner to absorb moisture. These are great general-purpose glove for light jobs in any industry.
- Catalog No:
- 32889-289
- 93000-964
- 32889-297
- 93000-967
- 93000-968
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable/Reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Latex/Latex-free:
- Latex-Free
- Latex-Free
- Latex-Free
- Latex-Free
- Latex-Free
- Powdered/Powder-free:
- Powder-Free
- Powder-Free
- Powder-Free
- Powder-Free
- Powder-Free