Order Entry
United States
ContactUsLinkComponent
 
Sodium thiosulphate concentrate for dilution to 0.1 N in aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®
Catalog # JT4695-1
Supplier:  Avantor
CAS Number:  
MOC:  Non-RegulatedCertificates
undefined
Sodium thiosulphate concentrate for dilution to 0.1 N in aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®
Catalog # JT4695-1
Supplier:  Avantor
CAS Number:  
MOC:  Non-RegulatedCertificates
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Pack type:
    LDPE ampoule
  • Size:
    50 ml
  • Environmentally Preferable:
  • Cat. No.:
    JT4695-1
  • Supplier no.:
    4695-01

Specifications

Specification Test Results

  • Normality (by titrimetry):
    Passes Test

Specification Test Results

About this item

Synonyms: Sodiumthiosulphate, Sodium hyposulphite, Antichlor, di-Sodium thiosulphate
 :
 :158.11 g/mol
Boiling Pt:100
Melting Pt:48
Storage Temperature:Ambient
EINECS:231-867-5
CAS Number:7772-98-7
Formula:Na₂S₂O₃
MDL Number:MFCD00003499
Merck Index:12,08844
 :Management of Change (MOC) category = N