Order Entry
Puerto Rico
ContactUsLinkComponent
Sodium hydroxide concentrate for dilution to 1 N in aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®
Corrosive
Sodium hydroxide concentrate for dilution to 1 N in aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®
Catalog # JT4689-1
CAS Number:  
MOC:  Non-RegulatedCertificates
undefined
Corrosive
Sodium hydroxide concentrate for dilution to 1 N in aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®
Catalog # JT4689-1
Supplier Number:  4689-01
CAS Number:  
MOC:  Non-RegulatedCertificates

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    LDPE ampoule, DILUT-IT®
  • Size:
    1 pack
  • Cat. No.:
    JT4689-1
  • Supplier no.:
    4689-01

Specifications

Specification Test Results

  • Normality (by titrimetry):
    Passes Test

Specification Test Results

About this item

 :
 :40 g/mol
Boiling Pt:∼105 °C (1013 hPa)
Density:1.327 g/cm³ (20 °C)
Storage Temperature:Ambient
ADR:8,II
 :Management of Change (MOC) category = N
EINECS:215-185-5
UN:1824
 :GHS05
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548