Sodium hydroxide ≥98% (by acidimetry), pellets, BAKER ANANYZED™, Electronic Grade for microelectronic, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIALS US
MOC: Regulated
About this item
Synonyms: Caustic soda
CAS Number:1310-73-2
:
Formula:NaOH
MDL Number:MFCD00003548
:40 g/mol
Merck Index:14,08627
Boiling Pt:1390 °C (1013 hPa)
Melting Pt:323 °C
Density:2.13 g/cm³ (20 °C)
UN:1823
ADR:8,II
:Management of Change (MOC) category = R
Storage Temperature:Ambient
EINECS:215-185-5
REACH:01-2119457892-27
:GHS05
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay (NaOH) (by acidimetry):≥98.0 %
- Sodium Carbonate (Na₂CO₃):≤0.4 %
- Insoluble Matter:≤30 ppm
- Ammonium Hydroxide Precipitate:≤0.01 %
- Potassium (K)(by FES):≤0.02 %
- Nitrogen Compounds (as N):≤3 ppm
- Chloride (Cl):≤0.005 %
- Phosphate (PO₄):≤2 ppm
- Sulfate (SO₄):≤30 ppm
- Trace Impurities - Copper (Cu):≤10 ppm
- Heavy Metals (as Ag):≤10 ppm
- Trace Impurities - Iron (Fe):≤3.000 ppm
- Trace Impurities - Mercury (Hg)(by AAS):≤0.100 ppm
- Trace Impurities - Nickel (Ni):≤5.000 ppm
Specification Test Results
Related Information
Certificates
Search by Lot Number3723 Product Specifications VWR GHS Compliance Letter (US)
Additional Product Details
Avantor Portfolio of Chemicals
Product Literature
Brand Definition Baker Analyzed