Order Entry
Puerto Rico
ContactUsLinkComponent
Sodium hydroxide 50% in aqueous solution, CMOS for the electronics industry, J.T.Baker®
Corrosive
Irritant
Sodium hydroxide 50% in aqueous solution, CMOS for the electronics industry, J.T.Baker®
Catalog # JT3719-09
CAS Number:  
undefined
Corrosive
Irritant
Sodium hydroxide 50% in aqueous solution, CMOS for the electronics industry, J.T.Baker®
Catalog # JT3719-09
Supplier Number:  3719-09
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Poly bottle
  • Size:
    600 lbs.
  • Cat. No.:
    JT3719-09

Specifications

Specification Test Results

  • Assay (NaOH) (by acidimetry):
    49.5 - 51.5 %
  • Color (APHA):
    ≤10
  • Insoluble Matter:
    ≤30 ppm
  • Sodium Carbonate (Na₂CO₃):
    ≤0.05 %
  • Ammonium Hydroxide Precipitate:
    ≤0.005 %
  • Chloride (Cl):
    ≤20 ppm
  • Heavy Metals (as Ag):
    ≤2 ppm
  • Nitrogen Compounds (as N):
    ≤1 ppm
  • Phosphate (PO₄):
    ≤1 ppm
  • Trace Impurities - Potassium (K):
    ≤50 ppm
  • ACS - Sulfate (SO₄):
    ≤5 ppm
  • Trace Impurities - Aluminum (Al):
    ≤1000.0 ppb
  • Trace Impurities - Calcium (Ca):
    ≤1500.0 ppb
  • Trace Impurities - Chromium (Cr):
    ≤500.0 ppb
  • Trace Impurities - Copper (Cu):
    ≤100.0 ppb
  • Trace Impurities - Iron (Fe):
    ≤1000.0 ppb
  • Trace Impurities - Lead (Pb):
    ≤300.0 ppb
  • Trace Impurities - Magnesium (Mg):
    ≤500.0 ppb
  • Trace Impurities - Mercury (Hg):
    ≤50 ppb
  • Trace Impurities - Nickel (Ni):
    ≤100.0 ppb
  • Trace Impurities - Silicon (Si):
    ≤2000.0 ppb
  • Trace Impurities - Zinc (Zn):
    ≤300.0 ppb

Specification Test Results

About this item

 :
 :40 g/mol
Boiling Pt:119…135 °C (1013 hPa)
Melting Pt:9…10 °C
Density:1.53 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-185-5
UN:1824
ADR:8,II
 :Management of Change (MOC) category = R
 :GHS05
 :GHS07
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548