


Hexachloroplatinic(IV) acid hexahydrate Pt ∼ 37,5%, BAKER ANALYZED® ACS
Catalog # JT2890-3
Supplier: AVANTOR PERFORMANCE MATERIALS US
CAS Number:
Specifications
- Pack type:Bottle
- Size:1 g
- Cat. No.:JT2890-3
- Supplier no.:2890-03
Specifications
Specification Test Results
- Solubility in Alcohol:Passes Test
- Assay (as Pt):≥37.50 %
- Alkalies and Other Salts (as SO₄):≤0.05 %
- Suitability for Potassium Determination:Passes Test
Specification Test Results
About this item
:
:517.9 g/mol
Merck Index:13,07609
Melting Pt:150 °C
Density:2.434 g/cm³ (20 °C)
EINECS:241-010-7
UN:2507
ADR:8,III
:Management of Change (MOC) category = N
:GHS06
:GHS05
:GHS08
CAS Number:18497-13-7
Formula:H₂PtCl₆·6H₂O
MDL Number:MFCD00149910