
Orthophosphoric acid ≥85%, CMOS for the electronics industry, J.T.Baker®
Catalog # JT0264-5
Supplier: AVANTOR PERFORMANCE MATERIALS US
CAS Number:
MOC: RegulatedCertificates
Specifications
- Pack type:Poly bottle
- Size:14 lbs.
- Cat. No.:JT0264-5
- Supplier no.:0264-05
Specifications
Specification Test Results
- Assay (H₃PO₄) (by acidimetry) :85.0 - 87.0 %
- Color (APHA) :≤ 10 5
- Specific Gravity at 60°/60°F :1.691 - 1.710
- Reducing Substances:Passes Test
- Volatile Acids (µeq/g) :≤ 0.16
- Chloride (Cl) :≤ 1 ppm
- Nitrate (NO₃):≤ 2 ppm
- Sulfate (SO₄) :≤ 12 ppm
- Trace Impurities - Aluminum (Al) :≤ 0.500 ppm
- Trace Impurities - Antimony (Sb) :≤ 10.000 ppm
- Trace Impurities - Arsenic (As) :≤ 0.050 ppm
- Trace Impurities - Calcium (Ca) :≤ 1.500 ppm
- Trace Impurities - Chromium (Cr) :≤ 0.200 ppm
- Trace Impurities - Cobalt (Co) :≤ 0.050 ppm
- Trace Impurities - Copper (Cu) :≤ 0.050 ppm
- Trace Impurities - Gold (Au) :≤ 0.300 ppm
- Trace Impurities - ACS - Heavy Metals (as Pb) :≤ 5 ppm
- Trace Impurities - Iron (Fe) :≤ 2.000 ppm
- Trace Impurities - Lead (Pb) :≤ 0.300 ppm
- Trace Impurities - Lithium (Li) :≤ 0.100 ppm
- Trace Impurities - Magnesium (Mg) :≤ 0.20 ppm
- Trace Impurities - Manganese (Mn) :≤ 0.100 ppm
- Trace Impurities - Nickel (Ni) :≤ 0.200 ppm
- Trace Impurities - Potassium (K) :≤ 1.500 ppm
- Trace Impurities - Sodium (Na) :≤ 2.500 ppm
- Trace Impurities - Strontium (Sr) :≤ 0.100 ppm
- Trace Impurities - Titanium (Ti) :≤ 0.300 ppm
- Trace Impurities - Zinc (Zn) :≤ 2.000 ppm
- Particle Count - 1.0 µm and greater :≤ 10 par/ml
- Storage Conditions:Store at temperatures above 22°C (72°F)
Specification Test Results
About this item
:
:98 g/mol
Melting Pt:28 °C
Density:1.71 to 1.87 g/cm³ (25 °C)
Storage Temperature:Ambient
ADR:8,III
:Management of Change (MOC) category = R
EINECS:231-633-2
UN:1805
:GHS05
CAS Number:7664-38-2
Formula:H₃PO₄
MDL Number:MFCD00011340