Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-CD34 Mouse Monoclonal Antibody [clone: 4H11] (mFluor™ Violet)
Anti-CD34 Mouse Monoclonal Antibody [clone: 4H11] (mFluor™ Violet)
Supplier:  AAT Bioquest
Certificates
undefined
Anti-CD34 Mouse Monoclonal Antibody [clone: 4H11] (mFluor™ Violet)
Supplier:  AAT Bioquest

Specifications

  • Catalog No:
  • 77021-296
  • 77024-192
  • Antigen Symbol:
  • CD34
  • CD34
  • Antigen Name:
  • CD34 antigen
  • CD34 antigen
  • Antibody Type:
  • Secondary
  • Secondary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • mFluor™ Violet
  • mFluor™ Violet
  • Clone:
  • 4H11
  • 4H11
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Isotype:
  • IgG1
  • IgG1
  • Purification:
  • Affinity purification
  • Affinity purification
  • Storage Temperature:
  • Refrigerated (2...8 °C); Minimize light exposure
  • Refrigerated (2...8 °C); Minimize light exposure
  • Shipping Temperature:
  • 20
  • 20

Specifications

Frequently Bought Together