Order Entry
Puerto Rico
ContactUsLinkComponent
Nitrogen Standard, 1 ml = 0.1 mg N, 100 ppm N as Nitrate (442.7 ppm NO₃⁻), Ricca Chemical Company
Nitrogen Standard, 1 ml = 0.1 mg N, 100 ppm N as Nitrate (442.7 ppm NO₃⁻), Ricca Chemical Company
Catalog # RC545716
Supplier:  Ricca Chemical
Small Business Enterprise
CAS Number:  
undefined
Nitrogen Standard, 1 ml = 0.1 mg N, 100 ppm N as Nitrate (442.7 ppm NO₃⁻), Ricca Chemical Company
Catalog # RC545716
Supplier:  Ricca Chemical
Supplier Number:  R5457000-500A
Small Business Enterprise
CAS Number:  

Specifications

  • Concentration:
    100 ppm
  • Description:
    Nitrogen-nitrate standard
  • Size:
    16 oz.
  • Cat. No.:
    RC545716
  • Contents:
    KNO₃ in H₂O/tr. CHCl₃
  • Supplier no.:
    R5457000-500A

Specifications

About this item

APHA for nitrate

100 ppm nitrogen.