Soil and Compost Dial Thermometers
Supplier: Heathrow Scientific
Certificates
About this item
Our dial thermometers are an excellent choice for many applications including soil and compost testing, food and beverage quality control, and asphalt/concrete testing.
- Dial diameter 76.2 mm (3")
The dial thermometer indicates temperature through the metal stem with attached circular scale. It does don’t require batteries, it is rugged, and readings can be adjusted with the calibration screw on the back.