Round-Bottom Three-Neck Flasks, Indented Morton Type, Ace Glass
Supplier: Ace Glass
Certificates
About this item
Round-bottom flasks with three necks and [ST] outer joints
Indents allow more complete agitation. The outer joints are tooled with reinforcement rings for added strength and stability. Necks are vertical unless noted otherwise. Heavy wall flasks feature walls that are approximately 30% heavier than standard flasks.
Ordering information: Vertical neck flasks can be ordered for angled neck capacities; contact your Avantor sales representative.