Specifications
- Cat. No.:62379-567
- W×H×D:5.4×4.4×11.1 cm (2⅛×1¾×4⅜")
- Accuracy:0.01%
- Weight:255 g (9 oz.)
Specifications
About this item
Timer performs reproducible stirring, heating, or pumping times.
- Traceable®
- Countdown and Time-of-Day modes
- Case is high-impact ABS plastic
Use for timed shutdown or start-up of apparatus during any test
A manual override function allows equipment to be switched on/off instantly without disturbing the program. Rechargeable NiCad battery maintains settings in the event of an electrical outage. Three-prong grounded receptacle relay controls equipment rated to 125 V, 15 A, 1800 W, ¹/₃ hp. A green LED indicates the on/off mode of attached equipment.
Programmable in any one-minute interval from 23 hours, 59 minutes to 1 minute. Countdown mode turns any piece of equipment on or off at the end of a set time period, which eliminates scheduling time-of-day settings and reduces programming time. Time-of-day mode permits programming of 14 a.m./p.m. clock settings for each day of the week, up to 196 on/off settings per week.