

Magnesium perchlorate, AR® ACS desiccant, Macron Fine Chemicals™
Catalog # MK503904
Supplier: AVANTOR PERFORMANCE MATERIALS US
CAS Number:
MOC: Non-RegulatedCertificates
Specifications
- Pack type:Glass bottle, amber, wide mouth
- Size:500 g
- Cat. No.:MK503904
- Supplier no.:5039-04
Specifications
Specification Test Results
- Loss on Drying at 190°C:≤8 %
- ACS - Suitability for Moisture Absorption:Passes Test
- ACS - Titrable Free Acid (meq/g):≤0.005
- ACS - Titrable Base (meq/g):≤0.025
Specification Test Results
About this item
:
:223.21 g/mol
Merck Index:13,05702
Melting Pt:251 °C
Density:2.21 g/cm³ (20 °C)
EINECS:233-108-3
UN:1475
ADR:5.1,II
:Management of Change (MOC) category = N
:GHS07
:GHS03
CAS Number:10034-81-8
Formula:Cl₂MgO₈
MDL Number:MFCD00011102