Specifications
- Pack type:Poly bottle
- Size:200 per bottle
- Cat. No.:EM-6501
- Supplier no.:6501-200TAB
Specifications
About this item
White, concave tablets
Each tablet makes 1X solution when dissolved in 100mL distilled or deionized water (1X=137mM sodium chloride, 2.7mM potassium chloride, and 10mM sodium phosphate dibasic/potassium phosphate monobasic buffer). If higher concentrations are desired, dissolve up to four tablets per 100mL water. DNase-, RNase-, and protease-free. Toxic effects.