Order Entry
Puerto Rico
ContactUsLinkComponent
Methyl ethyl ketone ≥99.5% (by GC), BAKER ANALYZED® HPLC for HPLC, UHPLC, for spectrophotometry, J.T.Baker®
Flammable
Irritant
Methyl ethyl ketone ≥99.5% (by GC), BAKER ANALYZED® HPLC for HPLC, UHPLC, for spectrophotometry, J.T.Baker®
Catalog # JT9214-3
   Safer Human and Environmental Health
CAS Number:  
MOC:  Non-RegulatedCertificates
undefined
Flammable
Irritant
Methyl ethyl ketone ≥99.5% (by GC), BAKER ANALYZED® HPLC for HPLC, UHPLC, for spectrophotometry, J.T.Baker®
Catalog # JT9214-3
Supplier Number:  9214-03
   Safer Human and Environmental Health
CAS Number:  
MOC:  Non-RegulatedCertificates

Specifications

  • Pack type:
    Glass bottle, amber, narrow mouth, PVC-coated
  • Size:
    4 L
  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental Certification:
    Chem21 solvent selection guide recommended by default and after discussion
  • Cat. No.:
    JT9214-3
  • Supplier no.:
    9214-03

Specifications

Specification Test Results

  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 400-350 nm:
    ≤0.01
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 340 nm:
    ≤0.07
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 335 nm:
    ≤0.40
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - UV Cut-off, nm:
    ≤330
  • Fluorescence Trace Impurities, measured as Quinine Base - at 450 nm Emission:
    ≤0.5 ppb
  • Fluorescence Trace Impurities, measured as Quinine Base - at Emission Maximum for Impurities:
    ≤1.0 ppb
  • Assay (CH₃COCH₂CH₃) (by GC):
    ≥99.5 %
  • Residue after Evaporation:
    ≤3.0 ppm
  • Water (by KF, volumetric):
    ≤0.03 %

Specification Test Results

About this item

 :
Merck Index:13,06097
Boiling Pt:79.6 °C (1013 hPa)
Melting Pt:–86 °C
Density:0.8054 g/cm³ (20 °C)
Flash Pt:–4 °C (closed cup)
ADR:3,II
 :Management of Change (MOC) category = N
 :72.11 g/mol
Storage Temperature:Ambient
EINECS:201-159-0
UN:1193
REACH:01-2119457290-43
 :GHS02
 :GHS07
CAS Number:78-93-3
Formula:C₂H₅COCH₃
MDL Number:MFCD00011648