Rollpruf® Cleanroom Gloves, AdvanTech, Mapa Professionnel
Supplier: MAPA
Certificates
About this item
ISO Class 4 (FED-STD-209E Class 10/M2.5) gloves provide product protection for hand tasks in cleanroom applications including assembly, testing, and inspection in the semiconductor, optical, aerospace, and electronics industries.
- Triple-polymer blend of natural rubber latex, nitrile, and neoprene
- Hand-specific
Eextractables, and nonvolatile residue (NVR). Color-coded cuff band prevents roll down. Powder-free. Textured bisque grip. Color: natural with color-coded cuff band.
Certifications: Meet CFR 21 170-199 for food and drug handling, and IEST-RPCC-0052 wet tests for particulate, total extractables, and contribution to corrosion.
Ordering information: Individual pairs packaged in sealed antistatic bags and supplied in two antistatic master poly bags of 100 pair each. Bulk packaging consists of four sealed poly bags, 50 singles each.
Specifications
- Catalog No:
- 32931-752
- 32931-774
- 32932-074
- 32931-796
- 32932-132
- 32932-096
- 32932-110
- 32931-730
- Ambidextrous/Hand-specific:
- Hand-Specific
- Hand-Specific
- Hand-Specific
- Hand-Specific
- Hand-Specific
- Hand-Specific
- Hand-Specific
- Hand-Specific