Isobutanol ≥99.0%, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIALS US
MOC: Non-Regulated
About this item
CAS Number:78-83-1
:
Formula:(CH₃)₂CHCH₂OH
MDL Number:MFCD00004740
:74.12 g/mol
Merck Index:13,05148
Boiling Pt:107.9 °C (1013 hPa)
Melting Pt:-108 °C
Density:0.802 g/cm³ (20 °C)
EINECS:201-148-0
UN:1212
ADR:3,III
:Management of Change (MOC) category = N
Flash Pt:28 °C
:GHS02
:GHS05
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental Certification:Chem21 Solvent selection guide recommended by default and after discussion
Specifications
Specification Test Results
- Assay ((CH₃)₂CHCH₂OH) (by GC, corrected for water):≥99.0 %
- Carbonyl Compounds - 2-Butanone:≤0.02 %
- Carbonyl Compounds - Butyraldehyde:≤0.01 %
- Color (APHA):≤10
- Residue after Evaporation:≤0.001 %
- Solubility in H₂O:Passes Test
- Titrable Acid (meq/g):≤0.0005
- Water (H₂O)(by Karl Fischer titrn):≤0.1 %
Specification Test Results
Related Information
Certificates
Search by Lot Number9044 Product Specifications VWR GHS Compliance Letter (US)
Additional Product Details
Avantor Portfolio of Chemicals Avantor Top Rated Guide
Product Literature
Brand Definition Baker Analyzed
Datasheet
spec_JT9044-3