Order Entry
Puerto Rico
ContactUsLinkComponent
Soil Hydrometer, Specific Gravity, ASTM 151H
Soil Hydrometer, Specific Gravity, ASTM 151H
Catalog # 76546-590
undefined
Soil Hydrometer, Specific Gravity, ASTM 151H
Catalog # 76546-590
Supplier Number:  67151H

Specifications

  • Cat. No.:
    76546-590
  • ASTM no.:
    151H
  • Range:
    0.995...1.038 nD
  • Subdivisions:
    0.001 nD

Specifications

About this item

This hydrometer measures the specific gravity of soil samples in suspension.

  • Calibrated for use at 20 °C (68 °F)

This hydrometer is often used with a sedimentation cylinder for soil analysis.

Certifications: Manufactured to the specifications of the American Society for Testing and Materials (ASTM).