Nitri Task Foam™ Supported Foam Nitrile Gloves, Honeywell Safety
Catalog # 89022-132
Supplier: Avantor
Specifications
- Glove color:Gray
- Glove size:2X-Large
- Ambidextrous/Hand-specific:Ambidextrous
- Cat. No.:89022-132
- Supplier no.:NFF13/11XXL
Specifications
About this item
For use in automotive assembly, general maintenance work, light assembly, manufacturing, and small part handling applications.
- Color-coded cuff trim assists in easy size identification
- Knit wrist
The nitrile palm foam coating provides resistance to snags and abrasions, and gives an excellent grip in oily applications. Open back design allows gloves to breathe for added comfort. Minimal shrinkage of nylon shell allows gloves to be laundered.