ContactUsLinkComponent
HyFlex® 11-100 Static Control Gloves, Ansell
HyFlex® 11-100 Static Control Gloves, Ansell
Certificates
undefined
HyFlex® 11-100 Static Control Gloves, Ansell
Specifications
  • Catalog No:
  • 47727-530
  • 47727-526
  • 47727-524
  • 47727-528
  • Ambidextrous/Hand-specific:
  • Hand-Specific
  • Hand-Specific
  • Hand-Specific
  • Hand-Specific
  • Disposable/Reusable:
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Liner Material:
  • Conductive Fiber/Nylon
  • Conductive Fiber/Nylon
  • Conductive Fiber/Nylon
  • Conductive Fiber/Nylon
  • Finish:
  • Palm Coated
  • Palm Coated
  • Palm Coated
  • Palm Coated
  • Cuff Style:
  • Knit Wrist
  • Knit Wrist
  • Knit Wrist
  • Knit Wrist
  • EN Norm:
  • 3131
  • 3131
  • 3131
  • 3131
Specifications