

Hydrochloric acid concentrate for dilution to 0.1 mol (0.1N), DILUT-IT® dissolution media concentrate, J.T.Baker®
Catalog # JTD011-0
Supplier: AVANTOR PERFORMANCE MATERIALS US
CAS Number:
MOC: Non-RegulatedCertificates
Specifications
- Pack type:Poly bottle
- Size:230.8 ml
- Cat. No.:JTD011-0
- Supplier no.:D011-00
Specifications
Specification Test Results
- Appearance:Passes Test
- Normality at 26x dilution:0.098 - 0.102
Specification Test Results
About this item
:
Density:1 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-595-7
UN:1789
ADR:8,II
:Management of Change (MOC) category = N
:36.46
:GHS05
:GHS07
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324