Order Entry
Puerto Rico
ContactUsLinkComponent
Hydrochloric acid 37.0 - 38.0%, Finyte®, J.T.Baker®
Corrosive
Irritant
Hydrochloric acid 37.0 - 38.0%, Finyte®, J.T.Baker®
Catalog # JT5800-5
CAS Number:  
undefined
Corrosive
Irritant
Hydrochloric acid 37.0 - 38.0%, Finyte®, J.T.Baker®
Catalog # JT5800-5
Supplier Number:  5800-05
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Poly bottle, pressure bottom
  • Size:
    10 lbs.
  • Cat. No.:
    JT5800-5
  • Supplier no.:
    5800-05

Specifications

Specification Test Results

  • Assay (as HCl) (by acid-base titrn):
    37.0 - 38.0 %
  • Color (APHA):
    ≤ 10
  • Residue after Ignition:
    ≤ 3 ppm
  • Extractable Organic Substances:
    ≤ 3 ppm
  • Bromide (Br):
    ≤ 0.005 %
  • Free Halogen (as Cl₂):
    Passes Test
  • Phosphate (PO₄):
    ≤ 0.03 ppm
  • Sulfate (SO₄):
    ≤ 0.3 ppm
  • Sulfite (SO₃):
    ≤ 0.4 ppm
  • Trace Impurities - Aluminum (Al):
    ≤ 10.0 ppb
  • Arsenic and Antimony (as As):
    ≤ 3.0 ppb
  • Trace Impurities - Barium (Ba):
    ≤ 5.0 ppb
  • Trace Impurities - Beryllium (Be):
    ≤ 1.0 ppb
  • Trace Impurities - Bismuth (Bi):
    ≤ 10.0 ppb
  • Trace Impurities - Boron (B):
    ≤ 10.0 ppb
  • Trace Impurities - Cadmium (Cd):
    ≤ 5.0 ppb
  • Trace Impurities - Calcium (Ca):
    ≤ 20 ppb
  • Trace Impurities - Chromium (Cr):
    ≤ 10.0 ppb
  • Trace Impurities - Cobalt (Co):
    ≤ 5.0 ppb
  • Trace Impurities - Copper (Cu):
    ≤ 5.0 ppb
  • Trace Impurities - Gallium (Ga):
    ≤ 5.0 ppb
  • Trace Impurities - Germanium (Ge):
    ≤ 10.0 ppb
  • Trace Impurities - Gold (Au):
    ≤ 5.0 ppb
  • Trace Impurities - Iron (Fe):
    ≤ 10.0 ppb
  • Trace Impurities - Lead (Pb):
    ≤ 10.0 ppb
  • Trace Impurities - Lithium (Li):
    ≤ 5.0 ppb
  • Trace Impurities - Magnesium (Mg):
    ≤ 10.0 ppb
  • Trace Impurities - Manganese (Mn):
    ≤ 5.0 ppb
  • Trace Impurities - Molybdenum (Mo):
    ≤ 5.0 ppb
  • Trace Impurities - Nickel (Ni):
    ≤ 10.0 ppb
  • Trace Impurities - Niobium (Nb):
    ≤ 1.0 ppb
  • Trace Impurities - Potassium (K):
    ≤ 10.0 ppb
  • Trace Impurities - Silver (Ag):
    ≤ 5.0 ppb
  • Trace Impurities - Sodium (Na):
    ≤ 10.0 ppb
  • Trace Impurities - Strontium (Sr):
    ≤ 5.0 ppb
  • Trace Impurities - Tantalum (Ta):
    ≤ 5.0 ppb
  • Trace Impurities - Thallium (Tl):
    ≤ 5.0 ppb
  • Trace Impurities - Tin (Sn):
    ≤ 5.0 ppb
  • Trace Impurities - Titanium (Ti):
    ≤ 5.0 ppb
  • Trace Impurities - Vanadium (V):
    ≤ 1.0 ppb
  • Trace Impurities - Zinc (Zn):
    ≤ 5.0 ppb
  • Trace Impurities - Zirconium (Zr):
    ≤ 5.0 ppb
  • Particle Count - 0.5 µm and greater:
    ≤ 25 par/ml
  • Particle Count - 1.0 µm and greater:
    ≤ 10 par/ml

Specification Test Results

About this item

 :
Boiling Pt:110 °C (1013 hPa)
Melting Pt:–30 °C
Density:1.18 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:1789
ADR:8,II
 :Management of Change (MOC) category = R
 :36.46 g/mol
EINECS:231-595-7
 :GHS05
 :GHS07
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324
Boiling Pt:110 °C (1013 hPa)
Melting Pt:-30 °C