TAE Buffer, Concentrate, OmniPur*. 10X.
Supplier: MilliporeSigma
Certificates
About this item
Liquid.
- DNase-, RNase-, and protease-free
Each liter of concentrate makes 10 L of 1X TAE (0.04 M TRIS, 0.04 M Acetate, 0.001 M EDTA).
Specifications
- Storage Temperature:15...30 °C