Hexane (mixture of isomers) ≥98.5%, OmniSolv® HR-GC for gas chromatography, Supelco®
Catalog # EM-HX0297-1
Supplier: MilliporeSigma
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Pack type:Glass bottle
- Size:4 L
- Cat. No.:EM-HX0297-1
- Supplier no.:HX0297-1
Specifications
Specification Test Results
- Assay (n-hexane):Report
- Assay (total aliphatic hydrocarbons99.9% min
- Assay (total hexane isomers + methylcyclopentane):98.5% min
- Appearance:Clear liquid, free from particulates
- Cap. ECD responsive substances as PCNB (peak):2ng/L max
- Cap. ECD responsive substances as PCNB (total):20ng/L max
- Cap. FID responsive substances as decane (peak):1µg/L max
- Cap. FID responsive substances as decane (total):10µg/L max
- Color (APHA):5 max
- Filtered through 0.2 µm filter:To pass test
- Infrared spectrum:Conforms
- Residue after evaporation:1ppm max
- Titrable acid:0.2µeq/g max
- Water (H2O):0.01% max
Specification Test Results
About this item
CAS Number:110-54-3
:
Formula:H₃C(CH₂)₄CH₃
:86.18 g/mol
Merck Index:13,04712
Boiling Pt:68…70 °C (1013 hPa)
Boiling Pt:68 - 70 °C (1013 hPa)
Melting Pt:–95 °C
Melting Pt:-95 °C
Density:0.66 g/cm³ (20 °C)
Flash Pt:–22 °C
EINECS:203-777-6
UN:1208
ADR:3,II
:GHS02
:GHS08
:GHS07
:GHS09
Storage Temperature:Ambient
:92112-69-1
MDL Number:MFCD02179311
Flash Pt:-22 °C