

Hydrofluoric acid 47 - 51%, ARISTAR® ULTRA, Ultrapure for trace metal analysis, VWR Chemicals BDH®
Supplier: BDH
Certificates
About this item
CAS Number:7664-39-3
:
Formula:HF
MDL Number:MFCD00011346
:20.01 g/mol
Storage Temperature:Ambient
EINECS:231-634-8
UN:1790
ADR:8,II
:GHS06
:GHS05
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specification Test Results
- Assay (HF, w/w):47 - 51%
- Total Sulfur (S):100 ppb (ng/g) max.
- Aluminum (Al):20 ppt (pg/g) max.
- Antimony (Sb):20 ppt (pg/g) max.
- Arsenic (As):50 ppt (pg/g) max.
- Barium (Ba):10 ppt (pg/g) max.
- Beryllium (Be):10 ppt (pg/g) max.
- Bismuth (Bi):10 ppt (pg/g) max.
- Boron (B):100 ppt (pg/g) max.
- Cadmium (Cd):10 ppt (pg/g) max.
- Calcium (Ca):10 ppt (pg/g) max.
- Cerium (Ce):10 ppt (pg/g) max.
- Cesium (Cs):10 ppt (pg/g) max.
- Chromium (Cr):10 ppt (pg/g) max.
- Cobalt (Co):10 ppt (pg/g) max.
- Copper (Cu):10 ppt (pg/g) max.
- Dysprosium (Dy):1 ppt (pg/g) max.
- Erbium (Er):1 ppt (pg/g) max.
- Europium (Eu):1 ppt (pg/g) max.
- Gadolinium (Gd):1 ppt (pg/g) max.
- Gallium (Ga):10 ppt (pg/g) max.
- Germanium (Ge):10 ppt (pg/g) max.
- Gold (Au):20 ppt (pg/g) max.
- Hafnium (Hf):10 ppt (pg/g) max.
- Holmium (Ho):1 ppt (pg/g) max.
- Indium (In):1 ppt (pg/g) max.
- Iron (Fe):10 ppt (pg/g) max.
- Lanthanum (La):10 ppt (pg/g) max.
- Lead (Pb):10 ppt (pg/g) max.
- Lithium (Li):10 ppt (pg/g) max.
- Lutetium (Lu):1 ppt (pg/g) max.
- Magnesium (Mg):10 ppt (pg/g) max.
- Manganese (Mn):10 ppt (pg/g) max.
- Mercury (Hg):50 ppt (pg/g) max.
- Molybdenum (Mo):10 ppt (pg/g) max.
- Neodymium (Nd):1 ppt (pg/g) max.
- Nickel (Ni):20 ppt (pg/g) max.
- Niobium (Nb):10 ppt (pg/g) max.
- Palladium (Pd):20 ppt (pg/g) max.
- Platinum (Pt):20 ppt (pg/g) max.
- Potassium (K):10 ppt (pg/g) max.
- Praseodymium (Pr):1 ppt (pg/g) max.
- Rhenium (Re):10 ppt (pg/g) max.
- Rhodium (Rh):20 ppt (pg/g) max.
- Rubidium (Rb):20 ppt (pg/g) max.
- Ruthenium (Ru):20 ppt (pg/g) max.
- Samarium (Sm):1 ppt (pg/g) max.
- Scandium (Sc):10 ppt (pg/g) max.
- Selenium (Se):Information only
- Silver (Ag):10 ppt (pg/g) max.
- Sodium (Na):10 ppt (pg/g) max.
- Strontium (Sr):10 ppt (pg/g) max.
- Tantalum (Ta):Information only
- Tellurium (Te):1 ppt (pg/g) max.
- Terbium (Tb):1 ppt (pg/g) max.
- Thallium (Tl):10 ppt (pg/g) max.
- Thorium (Th):1 ppt (pg/g) max.
- Thulium (Tm):1 ppt (pg/g) max.
- Tin (Sn):20 ppt (pg/g) max.
- Titanium (Ti):20 ppt (pg/g) max.
- Tungsten (W):20 ppt (pg/g) max.
- Uranium (U):1 ppt (pg/g) max.
- Vanadium (V):10 ppt (pg/g) max.
- Ytterbium (Yb):1 ppt (pg/g) max.
- Yttrium (Y):1 ppt (pg/g) max.
- Zinc (Zn):10 ppt (pg/g) max.
- Zirconium (Zr):10 ppt (pg/g) max.
Specification Test Results
Related Information
Certificates
Search by Lot Numberspec_87003-221 spec_87003-222 spec_87003-223
Product specification
VWR GHS Compliance Letter (US)