Order Entry
Puerto Rico
ContactUsLinkComponent
Giemsa's stain stock solution 8.0 g/L in glycerol/methanol (1:1) for hematology, J.T.Baker®
Flammable
Irritant
Health
Giemsa's stain stock solution 8.0 g/L in glycerol/methanol (1:1) for hematology, J.T.Baker®
Catalog # JTM708-1
CAS Number:  
MOC:  Non-Regulated, With InformationCertificates
undefined
Flammable
Irritant
Health
Giemsa's stain stock solution 8.0 g/L in glycerol/methanol (1:1) for hematology, J.T.Baker®
Catalog # JTM708-1
Supplier Number:  M708-01
CAS Number:  
MOC:  Non-Regulated, With InformationCertificates

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    PE bottle
  • Size:
    500 ml
  • Cat. No.:
    JTM708-1
  • Supplier no.:
    M708-01

Specifications

Specification Test Results

  • Suitability for Blood Cell Staining:
    Passes Test

Specification Test Results

About this item

 :
Density:0.99 g/cm³ (20 °C)
EINECS:257-438-2
ADR:3,II
 :Management of Change (MOC) category = NI
UN:1230
 :GHS02
 :GHS07
 :GHS08
Storage Temperature:Ambient
CAS Number:51811-82-6
MDL Number:MFCD00081642