Order Entry
Puerto Rico
ContactUsLinkComponent
Water with 0,05% (v/v) formic acid, LabReady™, Solvent Blend for HPLC, Burdick & Jackson™
Water with 0,05% (v/v) formic acid, LabReady™, Solvent Blend for HPLC, Burdick & Jackson™
Certificates
Water with 0,05% (v/v) formic acid, LabReady™, Solvent Blend for HPLC, Burdick & Jackson™
Certificates

Specification Test Results

  • Formic Acid (v/v):
    0.045 - 0.055%
  • UV Cutoff:
    <190 nm
  • Residue:
    < 1 mg/L
  • Color (APHA):
    < 10
  • HPLC Gradient @ 215 nm:
    < 0.020 AU
  • HPLC Gradient @ 254 nm:
    < 0.010 AU

Specification Test Results

Related Information