
Ethanol denatured ≥92.0%, OmniSolv® (denatured with 5% methanol), Supelco®
Supplier: THOMAS SCIENTIFIC INC MX
Certificates
About this item
CAS Number:64-17-5
:
Formula:H₃CCH₂OH
MDL Number:MFCD00003568
:46.07 g/mol
Boiling Pt:~ 78 °C (1013 hPa)
Density:0.8 g/cm³ (20 °C)
Flash Pt:13 °C
Storage Temperature:Ambient
EINECS:200-578-6
UN:1170
ADR:3,II
:GHS02
Specification Test Results
- Capillary ECD responsive substances (as PCNB):5ng/L max
- Color (APHA):10 max
- Filtered through 0.2 µm filter:To pass test
- Form:Clear liquid
- Infrared spectrum:Conforms to standard
- Water (H2O):0.5% max
Specification Test Results
Related Information
Certificates
Search by Lot NumberAYN4_Analytical_Chemistry_-_Chromatography MilliporeSigma The New M-Bottle for Solvents.pdf
Product Literature
ghs_EX0278
SDS
VWR GHS Compliance Letter (US)