

Alcohol ≥82-86%, Reagent Grade (denatured with methanol and isopropanol), Supelco®
Catalog # EM-AX0442-3
Supplier: MilliporeSigma
CAS Number:
Specifications
- Pack type:Plastic bottle
- Size:4 L
- Cat. No.:EM-AX0442-3
- Supplier no.:AX0442-3
Specifications
About this item
:
UN:1170
ADR:3,III
:GHS02
:GHS08
CAS Number:64-17-5
MDL Number:MFCD00003568