Order Entry
Puerto Rico
ContactUsLinkComponent
Ethanol denatured 94.0-96.0% (v/v), GR ACS for histology, for biochemistry (denatured with 5% isopropanol and 5% methanol), Supelco®
Flammable
Health
Irritant
Ethanol denatured 94.0-96.0% (v/v), GR ACS for histology, for biochemistry (denatured with 5% isopropanol and 5% methanol), Supelco®
Supplier:  MilliporeSigma
Certificates
Flammable
Health
Irritant
Ethanol denatured 94.0-96.0% (v/v), GR ACS for histology, for biochemistry (denatured with 5% isopropanol and 5% methanol), Supelco®
Supplier:  MilliporeSigma
Certificates

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specification Test Results

  • Assay (ethanol + methanol):
    94.0-96.0% (v/v)
  • Assay (ethanol):
    89.0-92.0% (v/v)
  • Assay (isopropyl alcohol):
    4.0-6.0% (v/v)
  • Assay (methanol):
    3.5-5.5% (v/v)
  • Color (APHA):
    10 max
  • Form:
    Clear liquid
  • Infrared spectrum:
    Conforms to standard
  • Residue after evaporation:
    0.001% max
  • Water (H2O):
    0.075% max

Specification Test Results

Frequently Bought Together