ContactUsLinkComponent
EnzyChrom™ Hexokinase Assay Kit
EnzyChrom™ Hexokinase Assay Kit
  77441-684
 :  
undefined
EnzyChrom™ Hexokinase Assay Kit
  77441-684
 :  EHXK-100
 :  
  • Description:
    EnzyChrom™ Hexokinase assay kit
  • Size:
    100 tests
  • Cat. No.:
    77441-684
Hexokinase activity determination in biological samples (e.g. microorganisms, plasma, serum, cultured cells, tissue and culture media).
  • Fast and sensitive
  • Linear detection range (20 μl sample): 0.22 to 100 U/L for 15 min reaction
  • Convenient
  • Homogeneous 'mix-incubate-measure' type assay
  • High-throughput
  • Can be readily automated on HTS liquid handling systems for processing thousands of samples per day
HEXOKINASE (E.C. 2.7.1.1) is a mitochondrial and cytosolic enzyme and the first committed step in the glycolysis pathway. Hexose sugars, mainly glucose, are phosphorylated using ATP to form glucose-6-phosphate. Hexokinases are implicated in a myriad of human diseases, including cancer and diabetes. BioAssay Systems’ non-radioactive, colorimetric Hexokinase assay involves a stepwise reaction based on the reduction of the tetrazolium salt MTT in an NADPH-coupled enzymatic reaction to a colored product which exhibits an absorption maximum at 565 nm. The increase in absorbance at 565 nm is proportional to the enzyme activity.