Order Entry
Puerto Rico
ContactUsLinkComponent
QuantiChrom™ Pyrophosphatase Assay Kit
QuantiChrom™ Pyrophosphatase Assay Kit
Catalog # 77441-668
Supplier:  BIOASSAY SYSTEMS
CAS Number:  
undefined
QuantiChrom™ Pyrophosphatase Assay Kit
Catalog # 77441-668
Supplier:  BIOASSAY SYSTEMS
Supplier Number:  DPPT-100
CAS Number:  

Specifications

  • Description:
    QuantiChrom™ Pyrophosphatase assay kit
  • Size:
    100 tests
  • Cat. No.:
    77441-668

Specifications

About this item

For detection and quantification of pyrophosphatase enzyme activity.

  • Safe and sensitive
  • Non-radioactive assay
  • Use as little as 10 μl samples
  • Linear detection range in 96-well plate: 1.0 to 20 U/L activity
  • Fast and convenient
  • The procedure involves addition of a single working reagent and incubation for 60 min
  • Room temperature assay
  • No 37 °C incubator is needed
  • High-throughput
  • Homogeneous 'mix-incubate-measure' type assay
  • Can be readily automated to assay thousands of samples per day

INORGANIC PYROPHOSPHATASE(E.C.3.6.1.1) catalyzes the hydrolysis of phosphoester bonds on inorganic pyrophosphate [P2O74-], thereby releasing two orthophosphate molecules. Family I PPases are essential enzymes found in all kingdoms of life and are responsible for maintaining the correct pyrophosphate equilibrium necessary to carry out nucleic acid and protein synthesis, and facilitate fatty acid β-oxidation. Simple, direct and automation-ready procedures for measuring pyrophosphatase activity are very desirable. BioAssay Systems’ DPPT-100 assay is based on our proprietary phosphate assay kit (POMG-25H). The color intensity, measured at 620 nm, is proportionate to the amount of phosphate released from pyrophosphate hydrolysis.