ContactUsLinkComponent
QuantiChrom™ β-Glucuronidase Assay Kit
Catalog # 77441-656
Supplier:  BIOASSAY SYSTEMS
CAS Number:  
undefined
QuantiChrom™ β-Glucuronidase Assay Kit
Catalog # 77441-656
Supplier:  BIOASSAY SYSTEMS
Supplier Number:  DGCD-100
CAS Number:  
Specifications
  • Description:
    QuantiChrom™ β-Glucuronidase assay kit
  • Size:
    100 tests
  • Cat. No.:
    77441-656
Specifications
About this item
For quantitative determination of ß-glucuronidase enzyme activity in tissue and bacterial samples.
  • Highly sensitive
  • The fluorimetric nature of the kit detects 1×10⁻⁴ to 8 U/L BG activity in 96 well plate with only 10 μl sample within 30 min
  • Versatile
  • This assay has been validated to work with both animal and E.coli derived β-glucuronidase
  • High-throughput
  • Homogeneous 'mix-incubate-measure' type assay
  • Can be readily automated to assay thousands of samples per day
BETA-GLUCURONIDASE (BG) is an enzyme which catalyzes the hydrolysis of β-glucuronide bonds. In humans, β-glucuronidase catalyzes the hydrolysis of β-D-glucuronic acids from glycoproteins and mucopolysaccharides such as heparan sulfate. β-glucuronidase activity is important for drug metabolism as drugs are often conjugated to glucuronic acid to make for a more water soluble delivery system, relying on subsequent β-glucuronidase activity to release the active drug. Bilirubin clearance is also facilitated by glucuronidation. GUSB is often used as a reporter gene to track gene expression through β-glucuronidase activity. BioAssay Systems’ fluorimetric BG assay uses a β-D-glucuronide substrate which fluoresces upon hydrolysis with β-glucuronidase and is read at 365/450 nm.