QuantiFluo™ Asparaginase Assay Kit
Catalog # 77441-652
Supplier: BIOASSAY SYSTEMS
CAS Number:
Specifications
- Description:QuantiFluo™ Asparaginase assay kit
- Size:100 tests
- Cat. No.:77441-652
Specifications
About this item
For quantitative determination of asparaginase activity in biological samples.
- Safe and fast
- Non-radioactive assay
- Assay can be completed within 40 minutes
- Sensitive and accurate
- Linear detection range 1.1 to 300 U/L asparaginase in a 96-well plate assay
- Convenient and high-throughput
- Homogeneous 'mix-incubate-measure' type assay
- Can be readily automated to assay thousands of samples per day
ASPARAGINASE (EC 3.5.1.1) is a key enzyme in the metabolism of asparagine. Its principal use is as treatment for acute lymphoblastic leukemia and lymphoblastic lymphoma where the asparagine is an essential amino acid. BioAssay Systems’ DASN-100 Kit provides a convenient fluorimetric method to measure asparaginase activity in biological samples. In this assay, o-phthalaldehyde reacts with liberated ammonia where the increase in fluorescence at λex/em = 415/475 nm is directly proportional to enzyme activity.