Order Entry
Puerto Rico
ContactUsLinkComponent
QuantiFluo™ Glutaminase Assay Kit
QuantiFluo™ Glutaminase Assay Kit
Catalog # 77441-658
Supplier:  BIOASSAY SYSTEMS
CAS Number:  
undefined
QuantiFluo™ Glutaminase Assay Kit
Catalog # 77441-658
Supplier:  BIOASSAY SYSTEMS
Supplier Number:  DGLN-100
CAS Number:  

Specifications

  • Description:
    QuantiFluo™ glutaminase assay kit
  • Size:
    100 tests
  • Cat. No.:
    77441-658

Specifications

About this item

For quantitative determination of glutaminase activity in biological samples.

  • Fast and safe
  • Assay can be completed within 40 minutes
  • Non-radioactive assay
  • Sensitive and accurate
  • Linear detection range is 0.66 to 500 U/L glutaminase in a 96-well plate assay
  • Convenient and high-throughput
  • Homogeneous 'mix-incubate-measure' type assay
  • Can be readily automated to assay thousands of samples per day

Glutaminase (EC 3.5.1.2) is a key enzyme in the metabolism of glutamine. Glutamine is an important amino acid in the metabolism of cancer cells, and increased glutaminolysis has been linked to cancer. BioAssay systems DGLN-100 kit provides a convenient fluorimetric method to measure glutaminase activity in biological samples.

In this assay, o-phthalaldehyde reacts with liberated ammonia where the increase in fluorescence at λex/em = 415/475 nm is directly proportional to enzyme activity.