ContactUsLinkComponent
QuantiChrom™ Acetoacetate Assay Kit
QuantiChrom™ Acetoacetate Assay Kit
Catalog # 77441-642
Supplier:  BIOASSAY SYSTEMS
CAS Number:  
undefined
QuantiChrom™ Acetoacetate Assay Kit
Catalog # 77441-642
Supplier:  BIOASSAY SYSTEMS
Supplier Number:  DAAT-100
CAS Number:  
Specifications
  • Description:
    QuantiChrom™ acetoacetate assay kit
  • Size:
    100 tests
  • Cat. No.:
    77441-642
Specifications
About this item
Direct assay of acetoacetate in serum, plasma, urine and other biological samples.
  • Sensitive and accurate
  • Linear detection range of 0.012 to 1 mM (1.2 to 100 nmole/well) for acetoacetate in 96-well plate assay
  • Convenient
  • No sample pretreatment needed
  • The procedure involves adding a single working reagent and reading the optical density at room temperature in kinetic mode
  • High-throughput
  • Can be automated as a high-throughput 96-well plate assay for thousands of samples per day
Acetoacetate is one of three ketone bodies produced in the liver mainly from oxidation of fatty acids and are normally present at low concentrations in urine and blood. Increased ketone concentrations in the blood may lead to metabolic acidosis, which has been associated with diabetes, childhood hypoglycemia, growth hormone deficiency, alcohol or salicylate intoxication and inborn errors of metabolism.
Acetoacetate is an unstable ketone body that spontaneously decarboxylates to release carbon dioxide and acetone. Simple, direct and automation-ready procedures for measuring acetoacetate (AAT) are very desirable. BioAssay Systems QuantiChrom™ acetoacetate assay is based on a reaction with sodium nitroprusside forming a product with absorbance at 550 nm.