Order Entry
Puerto Rico
ContactUsLinkComponent
Reagent alcohol, anhydrous ACS, meets analytical specification of USP for HPLC, for spectrophotometry (denatured with methanol and isopropanol), Burdick & Jackson™
Irritant
Health
Flammable
Reagent alcohol, anhydrous ACS, meets analytical specification of USP for HPLC, for spectrophotometry (denatured with methanol and isopropanol), Burdick & Jackson™
   Sustainable Options Available
Certificates
undefined
Irritant
Health
Flammable
Reagent alcohol, anhydrous ACS, meets analytical specification of USP for HPLC, for spectrophotometry (denatured with methanol and isopropanol), Burdick & Jackson™
   Sustainable Options Available
Certificates

Specifications

  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental Certification:
    EPA safer chemical solvent (Green circle rating); Chem21 solvent selection guide recommended by default and after discussion

Specifications

Specification Test Results

  • Color (APHA):
    10 max.
  • GC Assay (Ethanol + Methanol):
    94 - 96 %
  • GC Assay (Isopropyl Alcohol):
    4 - 6 %
  • Residue:
    1 mg/L max.
  • Specific Gravity (25ºC/25ºC):
    0.785 - 0.795
  • UV Cutoff:
    210 nm max.
  • Water by Karl Fischer Titration:
    0.2 % max.

Specification Test Results

Frequently Bought Together