Order Entry
Puerto Rico
ContactUsLinkComponent
Ethyl acetate ≥99.9%, B&J Brand™, meets UV Specifications for HPLC, for pesticide residue analysis, for spectrophotometry carbonyl-free, Burdick & Jackson™
 
 
Ethyl acetate ≥99.9%, B&J Brand™, meets UV Specifications for HPLC, for pesticide residue analysis, for spectrophotometry carbonyl-free, Burdick & Jackson™
  BJ099-4
Product available on GSA Advantage®
 :  
undefined
 
 
Ethyl acetate ≥99.9%, B&J Brand™, meets UV Specifications for HPLC, for pesticide residue analysis, for spectrophotometry carbonyl-free, Burdick & Jackson™
  BJ099-4
 :  099-4
Product available on GSA Advantage®
 :  

 

  • Pack type:
    Glass bottle
  • Size:
    4 L
  • Cat. No.:
    BJ099-4
  • Supplier no.:
    099-4

 

 

  • Color with Sulfuric Acid:
    Passes test
  • Water by Karl Fischer Titration:
    0.05 % max.
  • Electron Capture GC:
    10 ng/L max.
  • GC Analysis:
    99.9 % min.
  • Refractive Index (20ºC):
    1.3719 - 1.3727
  • Residue:
    1 mg/L max.
  • Substances Reducing Permanganate:
    Passes test
  • UV Absorbance @ 275 nm:
    0.005 AU max.
  • UV Absorbance @ 300 nm:
    0.005 AU max.
  • UV Cutoff:
    256 nm max.

 

 

 :
 :88.11 g/mol
 :13,03792
 :77 °C
 :77.1 °C (1013 hPa)
 :–84 °C
 :-83 °C
 :0.902 g/cm³ (20 °C)
 :–4 °C
 :-4 °C
 :Ambient
 :205-500-4
 :1173
 :3,II
 :01-2119475103-46
 :GHS07
 :GHS02
 :141-78-6
 :CH₃COOC₂H₅
 :MFCD00009171