Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

n-Decane ≥99.0%

 
 
n-Decane ≥99.0%
 
n-Decane ≥99.0%
n-Decane ≥99.0%

 

 
   
 

CAS Number: 124-18-5
MDL Number: MFCD00008954
Molecular Formula: C10H22
Molecular Weight: 142.29
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 174
Melting point (°C): -30
Flash Point (°C): 50
Specific Gravity (20/20): 0.73

  •  :124-18-5
  •  :H₃C(CH₂)₈CH₃
  •  :MFCD00008954
  •  :142.28 g/mol
  •  :174 °C
  •  :–30 °C
  •  :0.73 (20 °C)
  •  :50 °C
  •  :Ambient
  •  :204-686-4
  •  :2247
  •  :3,III
 

 

  • Purity/Analysis Method:
    >99.0% (GC)
Product Family Options

 

  • Pack type
    Size
     
     
  • 25 ml
       
       
    Each (25ml)
    Each (25ml) - $26.00
    $26.00
    Each (25ml)
       $26.00

     : D0011-25ML
  • Bottle
    500 ml
       
       
    Each (500ml)
    Each (500ml) - $110.00
    $110.00
    Each (500ml)
       $110.00

     : D0011-500ML

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.