Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
n-Decane

n-Decane

Glass bottle, amber, narrow mouth, 500 ml
Catalog # JTG143-7
Flammable
Health
Supplier #:  G143-07
n-Decane
Catalog # JTG143-7
CAS Number:  124-18-5
MOC:  Non-RegulatedCertificates
n-Decane
n-Decane

Glass bottle, amber, narrow mouth, 500 ml

CAS Number:  124-18-5
MOC:  Non-Regulated
Price
$259.46
Log in for availability
Each (500ml) Case of 12 (500ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
  • CAS Number:124-18-5
  • Formula:H₃C(CH₂)₈CH₃
  • MDL Number:MFCD00008954
  • Molecular Weight:142.28 g/mol
  • Boiling Pt:174.1 °C (1013 hPa)
  • Melting Pt:–30 °C
  • Density:0.73 g/cm³ (20 °C)
  • Flash Pt:46 °C
  • Storage Temperature:Ambient
  • EINECS:204-686-4
  • ADR:3,III
  •  :Management of Change (MOC) category = N
  • UN:2247
Specifications

Specification Test Results

  • Identification (by IR):
    Passes Test

Specifications

  • Pack type:
    Glass bottle, amber, narrow mouth
  • Size:
    500 ml

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.