

1,2-Dichlorobenzene ≥98.5%, J.T.Baker®
Catalog # JT9233-3
Supplier: Avantor
CAS Number:
MOC: Non-RegulatedCertificates
Specifications
- Pack type:Glass bottle, amber, narrow mouth
- Size:4 L
- Cat. No.:JT9233-3
- Supplier no.:9233-03
Specifications
Specification Test Results
- Assay (by GC, corrected for water):≥98.5 %
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 300 nm:≤0.30
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 305 nm:≤0.10
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 335 nm:≤0.05
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 375 nm:≤0.01
- Ultraviolet Absorbance (1.00-cm cell vs. water) - UV Cut-off, nm:≤296
- Residue after Evaporation:≤8 ppm
- Water (by Karl Fischer titrn):≤0.02 %
Specification Test Results
About this item
:
:147 g/mol
Merck Index:13,03081
Boiling Pt:180.5 °C (1013 hPa)
Melting Pt:–17 °C
Density:1.309 g/cm³ (20 °C)
Flash Pt:66 °C
EINECS:202-425-9
UN:1591
ADR:6.1,III
:Management of Change (MOC) category = N
:GHS07
:GHS09
Storage Temperature:Ambient
CAS Number:95-50-1
Formula:C₆H₄Cl₂
MDL Number:MFCD00000535