Dimethyl sulfoxide ≥99.5%, B&J Brand™ for HPLC, for gas chromatography, for spectrophotometry, Burdick & Jackson™
Supplier: HONEYWELL INTERNATIONAL INC MX
Certificates
About this item
CAS Number:67-68-5
:
Formula:(CH₃)₂SO
MDL Number:MFCD00002089
:78.14 g/mol
Merck Index:13,03285
Boiling Pt:189 °C
Melting Pt:18.5 °C
Density:1.104 g/cm³ (20 °C)
Flash Pt:28.5 %(V)
Storage Temperature:Ambient
EINECS:200-664-3
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental Certification:Chem21 Solvent selection guide recommended by default
Specifications
Specification Test Results
- GC Analysis:99.5 % min.
- Refractive Index (20ºC):1.4770 - 1.4800
- UV Absorbance @ 275 nm:0.5 AU max.
- UV Absorbance @ 300 nm:0.2 AU max.
- UV Absorbance @ 350 nm:0.02 AU max.
- UV Absorbance @ 400 nm:0.005 AU max.
- UV Cutoff:268 nm max.
- Water by Karl Fischer Titration:0.04 % max.